CO1H324A692A CombiPhos Catalysts Inc

2-Fluoro-6-(pyrimidin-5-yl)pyridine-3-boronic acid pinacol ester

  • Cas Number : 2223046-69-1
  • Molecular Weight : 301.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart