CO1H324A6936 CombiPhos Catalysts Inc

2-(Tetrahydrofuran-3-yloxy)thiazole-5-boronic acid pinacol ester

  • Cas Number : 1356469-05-0
  • Molecular Weight : 297.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart