CO1H324A6939 CombiPhos Catalysts Inc

N,N-dimethyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

  • Cas Number : 1310385-02-4
  • Molecular Weight : 248.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart