CO1H324A6951 CombiPhos Catalysts Inc

2-Cyano-6-(trifluoromethyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223035-07-0
  • Molecular Weight : 298.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart