CO1H324A6953 CombiPhos Catalysts Inc

2,6-BIS(TERT-BUTYL)PYRIDINE-4-BORONIC ACID PINACOL ESTER

  • Cas Number : 1392146-23-4
  • Molecular Weight : 317.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart