CO1H324A6957 CombiPhos Catalysts Inc

4-Fluoro-3-methylpyridine-5-boronic acid pinacol ester

  • Cas Number : 2223049-19-0
  • Molecular Weight : 237.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart