CO1H324A695A CombiPhos Catalysts Inc

N-ethyl-N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazol-2-amine

  • Cas Number : 1310405-17-4
  • Molecular Weight : 268.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart