CO1H324A6963 CombiPhos Catalysts Inc

2-(3-methyl-1H-pyrazol-1-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine

  • Cas Number : 1402240-38-3
  • Molecular Weight : 286.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart