CO1H324A6969 CombiPhos Catalysts Inc

N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazol-2-amine

  • Cas Number : 1416367-38-8
  • Molecular Weight : 254.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart