CO1H324A6976 CombiPhos Catalysts Inc

3-(Cyclopropyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223051-98-5
  • Molecular Weight : 245.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart