CO1H324A6977 CombiPhos Catalysts Inc

3-(Cyclobutyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223012-94-8
  • Molecular Weight : 259.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart