CO1H324A697D CombiPhos Catalysts Inc

3-Phenylpyridine-2-boronic acid pinacol ester

  • Cas Number : 2223012-78-8
  • Molecular Weight : 281.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart