CO1H324A697E CombiPhos Catalysts Inc

3-(2-Tolyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223035-18-3
  • Molecular Weight : 295.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart