CO1H324A6981 CombiPhos Catalysts Inc

3-(2-Fluorophenyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223036-10-8
  • Molecular Weight : 299.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart