CO1H324A6986 CombiPhos Catalysts Inc

1-(6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazin-2-yl)pyridin-2(1H)-one

  • Cas Number : 1402172-73-9
  • Molecular Weight : 299.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart