CO1H324A6989 CombiPhos Catalysts Inc

3-(3-Trifluoromethylphenyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223012-26-6
  • Molecular Weight : 349.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart