CO1H324A6995 CombiPhos Catalysts Inc

3-(Pyridin-3-yl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223012-22-2
  • Molecular Weight : 282.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart