CO1H324A699F CombiPhos Catalysts Inc

5-(TERT-BUTYL)PYRIDINE-3-BORONIC ACID PINACOL ESTER

  • Cas Number : 1627723-04-9
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart