CO1H324A69A2 CombiPhos Catalysts Inc

6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazin-2-amine

  • Cas Number : 1356108-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart