CO1H324A69A4 CombiPhos Catalysts Inc

3-(Ethoxy)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223013-08-7
  • Molecular Weight : 249.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart