CO1H324A69A6 CombiPhos Catalysts Inc

3-(Cyclobutoxy)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223049-44-1
  • Molecular Weight : 275.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart