CO1H324A69A9 CombiPhos Catalysts Inc

3-(tert-Butoxy)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223047-07-0
  • Molecular Weight : 277.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart