CO1H324A69B3 CombiPhos Catalysts Inc

3-(Cyclopentyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223042-56-4
  • Molecular Weight : 273.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Inchi Code 1S/C16H24BNO2/c1-15(2)16(3,4)20-17(19-15)14-9-10-18-11-13(14)12-7-5-6-8-12/h9-12H,5-8H2,1-4H3
Inchi Key KALVEYWISPVTJW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart