CO1H324A69B4 CombiPhos Catalysts Inc

3-(Cyclohexyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 1402240-83-8
  • Molecular Weight : 287.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclohexyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Inchi Code 1S/C17H26BNO2/c1-16(2)17(3,4)21-18(20-16)15-10-11-19-12-14(15)13-8-6-5-7-9-13/h10-13H,5-9H2,1-4H3
Inchi Key BKCJFAKOOGRMPU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart