CO1H324A69B9 CombiPhos Catalysts Inc

4-Bromo-5-(iso-propyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 1402233-27-5
  • Molecular Weight : 326.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart