CO1H324A69BE CombiPhos Catalysts Inc

2-Amino-5-bromopyridine-4-boronic acid pinacol ester

  • Cas Number : 2223027-23-2
  • Molecular Weight : 298.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart