CO1H324A69BF CombiPhos Catalysts Inc

4,6-Dibromopyridine-2-boronic acid pinacol ester

  • Cas Number : 1402237-94-8
  • Molecular Weight : 362.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart