CO1H324A69C1 CombiPhos Catalysts Inc

4-Chloro-5-(cyclopentyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223050-87-9
  • Molecular Weight : 307.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart