CO1H324A69C4 CombiPhos Catalysts Inc

4-Bromo-6-ethylpyridine-2-boronic acid pinacol ester

  • Cas Number : 1402226-87-2
  • Molecular Weight : 312.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart