CO1H324A69D3 CombiPhos Catalysts Inc

4-CHLORO-5-METHYLPYRIDINE-3-BORONIC ACID PINACOL ESTER

  • Cas Number : 2223012-31-3
  • Molecular Weight : 253.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart