CO1H324A69D7 CombiPhos Catalysts Inc

4-Amino-6-methylpyridine-3-boronic acid pinacol ester

  • Cas Number : 1668475-78-2
  • Molecular Weight : 234.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart