CO1H324A69DF CombiPhos Catalysts Inc

2-(tert-butyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-4-amine

  • Cas Number : 2223027-21-0
  • Molecular Weight : 276.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart