CO1H324A69ED CombiPhos Catalysts Inc

2-Bromo-5-(n-propyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 1402233-25-3
  • Molecular Weight : 326.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart