CO1H324A69EE CombiPhos Catalysts Inc

2-Amino-5-(n-propyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223050-17-5
  • Molecular Weight : 262.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart