CO1H324A69F3 CombiPhos Catalysts Inc

2-Bromo-5-cyclobutylpyridine-4-boronic acid pinacol ester

  • Cas Number : 1402233-42-4
  • Molecular Weight : 338.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart