CO1H324A6A05 CombiPhos Catalysts Inc

2-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyridazin-3(2H)-one

  • Cas Number : 2223053-53-8
  • Molecular Weight : 298.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart