CO1H324A6A0C CombiPhos Catalysts Inc

2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazol-2-yl)pyridazin-3(2H)-one

  • Cas Number : 2096333-22-9
  • Molecular Weight : 305.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart