CO1H324A6A0F CombiPhos Catalysts Inc

1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazin-2-yl)piperidin-2-one

  • Cas Number : 2223052-43-3
  • Molecular Weight : 303.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart