CO1H324A6A18 CombiPhos Catalysts Inc

5-Bromo-4-(cyclohexyl)pyridine-2-boronic acid pinacol ester

  • Cas Number : 2223012-41-5
  • Molecular Weight : 366.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart