CO1H324A6A1E CombiPhos Catalysts Inc

2-(4-methyl-1H-imidazol-1-yl)-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazine

  • Cas Number : 1402240-39-4
  • Molecular Weight : 286.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart