CO1H324A6A27 CombiPhos Catalysts Inc

1-(5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazol-2-yl)pyrrolidin-2-one

  • Cas Number : 1356495-87-8
  • Molecular Weight : 294.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart