CO1H324A6A2A CombiPhos Catalysts Inc

3-Bromo-2-(cyclobutyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223036-30-2
  • Molecular Weight : 338.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart