CO1H324A6A33 CombiPhos Catalysts Inc

3-Amino-6-(cyclopentyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223045-02-9
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart