CO1H324A6A35 CombiPhos Catalysts Inc

3-Amino-6-(tert-butyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223036-32-4
  • Molecular Weight : 276.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart