CO1H324A6A37 CombiPhos Catalysts Inc

3-Amino-6-(piperidino)pyridine-4-boronic acid pinacol ester

  • Cas Number : 2223012-79-9
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart