CO1H324A6A6E CombiPhos Catalysts Inc

2-(4-methyl-1H-imidazol-1-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • Cas Number : 1351231-76-9
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart