CO1H324A6A73 CombiPhos Catalysts Inc

2-(3-Methylpiperidin-1-yl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 1350748-07-0
  • Molecular Weight : 302.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart