CO1H324A6A83 CombiPhos Catalysts Inc

2-(2-Amino-5-pyrimidyl)pyridine-4-boronic acid pinacol ester

  • Cas Number : 1402172-69-3
  • Molecular Weight : 298.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart