CO1H324A6AA6 CombiPhos Catalysts Inc

1-methyl-4-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-4-yl)piperazine

  • Cas Number : 1402174-19-9
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart