CO1H324A6AB7 CombiPhos Catalysts Inc

2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4,4'-bipyridine

  • Cas Number : 2223049-83-8
  • Molecular Weight : 282.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart